Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML2A
Entrez Gene ID
102183077
Gene Name
olfactomedin like 2A
Gene Aliases
-
Chromosome Location
Chr.11: 95296800 - 95324120 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055754.1 XP_017911243.1 7 - 8 1479 84
XM_018055755.1 XP_017911244.1 7 - 8 1340 84

View Full Product Details