Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCL1A
Entrez Gene ID
102187352
Gene Name
T cell leukemia/lymphoma 1A
Gene Aliases
-
Chromosome Location
Chr.21: 59889760 - 59893482 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065931.1 XP_017921420.1 1 - 2 272 60

View Full Product Details