Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102186846
Entrez Gene ID
102186846
Gene Name
zinc finger CCHC domain-containing protein 16
Gene Aliases
ZCCHC16
Chromosome Location
Chr.X|NW_017189517: 31527271 - 31551060 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044092.1 XP_017899581.1 3 - 3 385 101

View Full Product Details