Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TM4SF18
Entrez Gene ID
102171741
Gene Name
transmembrane 4 L six family member 18
Gene Aliases
-
Chromosome Location
Chr.1: 118427710 - 118451683 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675458.3 XP_005675515.2 3 - 4 548 69
XM_005675459.3 XP_005675516.2 3 - 4 548 69

View Full Product Details