Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf29
Entrez Gene ID
102170583
Gene Name
chromosome 5 C12orf29 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 17613588 - 17624801 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679786.3 XP_005679843.1 4 - 5 616 75

View Full Product Details