Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC5A
Entrez Gene ID
102174546
Gene Name
G protein-coupled receptor class C group 5 member A
Gene Aliases
-
Chromosome Location
Chr.5: 95590388 - 95610786 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680813.3 XP_005680870.1 3 - 4 1231 68
GenBank mRNA JO349369.2 - 2 - 3 991 68

View Full Product Details