Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSI
Entrez Gene ID
102173741
Gene Name
arylsulfatase family member I
Gene Aliases
-
Chromosome Location
Chr.7: 48771934 - 48779193 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683352.3 XP_005683409.2 1 - 2 893 55
GenBank mRNA JT659895.1 - 2 - 3 1478 55

View Full Product Details