Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDI1
Entrez Gene ID
102168367
Gene Name
DNA damage inducible 1 homolog 1
Gene Aliases
-
Chromosome Location
Chr.15: 78053056 - 78055402 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689353.2 XP_005689410.2 1 - 1 1613 75

View Full Product Details