Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf49
Entrez Gene ID
102170896
Gene Name
chromosome 15 C11orf49 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 5923951 - 6135205 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690159.3 XP_005690216.1 5 - 6 589 106
XM_005690160.3 XP_005690217.1 5 - 6 593 106
XM_013969851.2 XP_013825305.1 5 - 6 593 106
GenBank mRNA JO630283.2 - 4 - 5 739 106

View Full Product Details