Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC2
Entrez Gene ID
102170011
Gene Name
doublecortin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 231699 - 369310 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696786.3 XP_005696843.1 2 - 3 473 84
GenBank mRNA JO433239.2 - 8 - 9 1082 84

View Full Product Details