Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMERB1
Entrez Gene ID
102173524
Gene Name
bMERB domain containing 1
Gene Aliases
C25H16orf45
Chromosome Location
Chr.25: 13735032 - 13881094 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697522.2 XP_005697579.2 4 - 5 585 70
GenBank mRNA JO634954.2 - 2 - 3 1391 70

View Full Product Details