Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARV1
Entrez Gene ID
100861106
Gene Name
ARV1 homolog, fatty acid homeostasis modulator
Gene Aliases
-
Chromosome Location
Chr.28: 41581807 - 41603103 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698968.3 XP_005699025.2 2 - 3 380 93
GenBank mRNA JO578817.1 - 2 - 3 446 93
KA320701.1 - 4 - 5 924 93

View Full Product Details