Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDNF
Entrez Gene ID
102176339
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.13: 28788241 - 28807531 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057133.1 XP_017912622.1 2 - 3 315 103
GenBank mRNA JO387324.2 - 2 - 3 603 103

View Full Product Details