Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXPH2
Entrez Gene ID
102172848
Gene Name
neurexophilin 2
Gene Aliases
-
Chromosome Location
Chr.2: 77754450 - 77881930 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO454370.2 - 1 - 2 405 93

View Full Product Details