Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB14
Entrez Gene ID
102169230
Gene Name
ankyrin repeat and SOCS box containing 14
Gene Aliases
-
Chromosome Location
Chr.22: 43755924 - 43771786 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973561.2 XP_013829015.2 9 - 10 1792 91

View Full Product Details