Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC108635630
Entrez Gene ID
108635630
Gene Name
CD48 antigen-like
Gene Aliases
-
Chromosome Location
Chr.3: 111386424 - 111399330 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046542.1 XP_017902031.1 2 - 3 380 98

View Full Product Details