Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXNL1
Entrez Gene ID
102174012
Gene Name
nucleoredoxin like 1
Gene Aliases
-
Chromosome Location
Chr.7: 103122649 - 103126883 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051707.1 XP_017907196.1 1 - 2 325 60

View Full Product Details