Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POP5
Entrez Gene ID
102173679
Gene Name
POP5 homolog, ribonuclease P/MRP subunit
Gene Aliases
-
Chromosome Location
Chr.17: 8273709 - 8275949 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691570.3 XP_005691627.2 3 - 4 401 72
GenBank mRNA KA327444.1 - 3 - 4 357 72

View Full Product Details