Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
R3HCC1
Entrez Gene ID
102181123
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 70212830 - 70221176 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052332.1 XP_017907821.1 4 - 5 1059 81
GenBank mRNA JO396282.2 - 4 - 5 628 81

View Full Product Details