Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IFITM10
Entrez Gene ID
102177600
Gene Name
interferon induced transmembrane protein 10
Gene Aliases
-
Chromosome Location
Chr.29: 50737075 - 50748980 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042774.1 XP_017898263.1 2 - 3 496 77
GenBank mRNA JO640797.2 - 2 - 3 735 77
JO640798.2 - 2 - 3 735 77

View Full Product Details