Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCLRE1B
Entrez Gene ID
102168405
Gene Name
DNA cross-link repair 1B
Gene Aliases
-
Chromosome Location
Chr.3: 91114070 - 91122342 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677854.3 XP_005677911.1 3 - 4 944 69

View Full Product Details