Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOMM22
Entrez Gene ID
102182677
Gene Name
translocase of outer mitochondrial membrane 22
Gene Aliases
-
Chromosome Location
Chr.5: 109239252 - 109241949 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681079.3 XP_005681136.1 3 - 4 403 96

View Full Product Details