Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WDR25
Entrez Gene ID
102179891
Gene Name
WD repeat domain 25
Gene Aliases
-
Chromosome Location
Chr.21: 64544299 - 64688258 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066203.1 XP_017921692.1 5 - 6 1372 70

View Full Product Details