Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKOR2
Entrez Gene ID
102182651
Gene Name
SKI family transcriptional corepressor 2
Gene Aliases
-
Chromosome Location
Chr.24: 47100547 - 47134782 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039371.1 XP_017894860.1 3 - 4 2732 88

View Full Product Details