Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM11
Entrez Gene ID
102179596
Gene Name
RNA binding motif protein 11
Gene Aliases
-
Chromosome Location
Chr.1: 21539934 - 21569086 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674770.3 XP_005674827.1 4 - 5 499 103
XM_013967692.2 XP_013823146.1 5 - 6 532 103
XM_018053315.1 XP_017908804.1 4 - 5 850 103

View Full Product Details