Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF15
Entrez Gene ID
102178448
Gene Name
TNF superfamily member 15
Gene Aliases
-
Chromosome Location
Chr.8: 103667338 - 103688934 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684350.3 XP_005684407.2 3 - 4 377 94

View Full Product Details