Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
102186058
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.21: 47218293 - 47421515 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065926.1 XP_017921415.1 10 - 11 2041 107

View Full Product Details