Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C23H6orf89
- Entrez Gene ID
-
102178865
- Gene Name
- chromosome 23 C6orf89 homolog
- Gene Aliases
-
-
- Chromosome Location
-
Chr.23: 37913948 - 37952739 on Build ARS1
- Gene Symbol
- CRYZL1
- Entrez Gene ID
-
102171097
- Gene Name
- crystallin zeta like 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 334139 - 369177 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO501164.2 |
- |
- |
1209 |
80 |
View Full Product Details