Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRD1
Entrez Gene ID
102178325
Gene Name
leucine rich repeats and death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 111094639 - 111113193 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678910.3 XP_005678967.2 4 - 5 2497 69

View Full Product Details