Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAM1
Entrez Gene ID
102177707
Gene Name
PML-RARA regulated adaptor molecule 1
Gene Aliases
-
Chromosome Location
Chr.7: 93317011 - 93324791 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013965219.2 XP_013820673.2 7 - 8 4231 60
GenBank mRNA JO324065.2 - 7 - 8 1227 60

View Full Product Details