Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSGN1
Entrez Gene ID
102177988
Gene Name
mesogenin 1
Gene Aliases
-
Chromosome Location
Chr.11: 80520591 - 80521166 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686983.1 XP_005687040.1 1 - 1 420 63

View Full Product Details