Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZAR1L
Entrez Gene ID
102182140
Gene Name
zygote arrest 1 like
Gene Aliases
-
Chromosome Location
Chr.12: 58019212 - 58027408 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687528.3 XP_005687585.2 2 - 3 1154 104

View Full Product Details