Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A31
Entrez Gene ID
102170261
Gene Name
solute carrier family 22 member 31
Gene Aliases
-
Chromosome Location
Chr.18: 15682551 - 15687166 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061879.1 XP_017917368.1 8 - 9 1337 96
XM_018061880.1 XP_017917369.1 8 - 9 1247 96
XM_018061881.1 XP_017917370.1 7 - 8 1181 96
GenBank mRNA KA326469.1 - 8 - 9 1348 96

View Full Product Details