Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPA3
Entrez Gene ID
102171909
Gene Name
OPA3 outer mitochondrial membrane lipid metabolism regulator
Gene Aliases
-
Chromosome Location
Chr.18: 54170904 - 54238712 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692642.3 XP_005692699.1 1 - 2 215 83

View Full Product Details