Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL27
Entrez Gene ID
102172191
Gene Name
ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.19: 42699567 - 42702237 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693883.3 XP_005693940.1 4 - 5 476 76
XM_005693885.3 XP_005693942.1 4 - 5 573 76
GenBank mRNA JO571040.2 - 4 - 5 432 76
KA320940.1 - 1 - 2 187 76

View Full Product Details