Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC61G
Entrez Gene ID
102171662
Gene Name
Sec61 translocon gamma subunit
Gene Aliases
-
Chromosome Location
Chr.22: 1304123 - 1310904 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695496.3 XP_005695553.1 3 - 4 487 138
GenBank mRNA JO580273.1 - 3 - 4 537 139

View Full Product Details