Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBP3
Entrez Gene ID
102177960
Gene Name
retinol binding protein 3
Gene Aliases
-
Chromosome Location
Chr.28: 3494828 - 3504268 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699333.2 XP_005699390.1 1 - 2 3050 90

View Full Product Details