Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTX3
Entrez Gene ID
102179601
Gene Name
pentraxin 3
Gene Aliases
-
Chromosome Location
Chr.1: 109705011 - 109711073 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048475.1 XP_017903964.1 2 - 3 681 107
GenBank mRNA KA307807.1 - 1 - 2 1028 107

View Full Product Details