Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8H9orf43
Entrez Gene ID
102172811
Gene Name
chromosome 8 C9orf43 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 102368472 - 102381353 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052630.1 XP_017908119.1 3 - 4 392 93

View Full Product Details