Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf54
Entrez Gene ID
102184035
Gene Name
chromosome 5 C12orf54 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 31170729 - 31195784 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680023.3 XP_005680080.2 4 - 5 344 96

View Full Product Details