Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAPC5
Entrez Gene ID
102189590
Gene Name
small nuclear RNA activating complex polypeptide 5
Gene Aliases
-
Chromosome Location
Chr.10: 87915572 - 87940637 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685128.3 XP_005685185.1 3 - 4 384 111

View Full Product Details