Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB1
Entrez Gene ID
102186994
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
-
Chromosome Location
Chr.19: 26933757 - 26944480 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693503.2 XP_005693560.2 3 - 4 308 65
XM_013972198.2 XP_013827652.2 3 - 4 293 65
XM_013972199.2 XP_013827653.1 3 - 4 293 65
XM_018064583.1 XP_017920072.1 3 - 4 293 65
GenBank mRNA KA311468.1 - 3 - 4 201 65

View Full Product Details