Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TESMIN
Entrez Gene ID
102186666
Gene Name
testis expressed metallothionein like protein
Gene Aliases
-
Chromosome Location
Chr.29: 47153525 - 47194020 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042745.1 XP_017898234.1 2 - 3 580 73

View Full Product Details