Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
RTN4RL2
Entrez Gene ID
106502923
Gene Name
reticulon 4 receptor like 2
Gene Aliases
-
Chromosome Location
Chr.15: 2551885 - 2567713 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059067.1 XP_017914556.1 2 - 3 857 63

View Full Product Details