Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C24H18orf21
Entrez Gene ID
102184393
Gene Name
chromosome 24 C18orf21 homolog
Gene Aliases
-
Chromosome Location
Chr.24: 21447763 - 21456917 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039772.1 XP_017895261.1 2 - 3 198 87

View Full Product Details