Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM42
Entrez Gene ID
102183367
Gene Name
tripartite motif containing 42
Gene Aliases
-
Chromosome Location
Chr.1: 127775598 - 127801959 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675503.2 XP_005675560.2 3 - 4 1859 76

View Full Product Details