Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3H1orf87
Entrez Gene ID
102190285
Gene Name
chromosome 3 C1orf87 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 35039660 - 35123565 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678339.3 XP_005678396.2 3 - 4 488 66

View Full Product Details