Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8H9orf85
Entrez Gene ID
102170318
Gene Name
chromosome 8 C9orf85 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 47901640 - 47965409 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683750.3 XP_005683807.1 3 - 4 436 106
GenBank mRNA JO457097.2 - 1 - 2 120 106
JO584023.1 - 3 - 4 469 106

View Full Product Details