Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H16orf86
Entrez Gene ID
102186511
Gene Name
chromosome 18 C16orf86 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 36481070 - 36482792 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692152.3 XP_005692209.1 1 - 2 124 82
XM_005692153.2 XP_005692210.1 1 - 2 209 82

View Full Product Details