Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM16
Entrez Gene ID
102185685
Gene Name
tripartite motif containing 16
Gene Aliases
-
Chromosome Location
Chr.19: 32710749 - 32726721 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693592.3 XP_005693649.2 3 - 4 1049 98
GenBank mRNA KA351406.1 - 3 - 4 858 98

View Full Product Details